Would you like to log in again?
Bruns Bone Curette, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
40-7228 40-7222 40-7203 40-7230 40-7205 40-7223 40-7336 40-7208 40-7331 40-7333 40-7221 40-7209 40-7206 40-7338 40-7229 40-7330 40-7225 40-7210 40-7335 40-7337 40-7332 40-7226 40-7207 40-7224 40-7334 40-7220 40-7202 40-7227
10196-740EA 188.39 USD InStock
10196-740 10196-742 10196-720 10196-726 10196-802 10196-728 10196-804 10196-722 10196-724 10196-800 10196-718 10196-212 10196-696 10196-772 10196-698 10196-774 10196-730 10196-210 10196-770 10196-736 10196-738 10196-716 10196-216 10196-810 10196-732 10196-734 10196-806 10196-808
Bruns Bone Curette, OR Grade, Sklar
Curettes
Bruns Bone Curette is a debulking instrument commonly used in orthopedic procedures and neurosurgery.
It is used to scrape away tissue and bone from the spine, bones, and joints. It is available in multiple sizes with various curvatures and tip styles to suit the surgical site and specifics of the procedure.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty