Would you like to log in again?
Love-Gruenwald Intervertebral Disc Rongeur, OR Grade, Sklar®
Supplier: Sklar
42-3160 42-3152 42-3158 42-3156 42-3150
10196-950EA 837.04 USD InStock
10196-950 10196-956 10196-958 10196-952 10196-954
Love-Gruenwald Intervertebral Disc Rongeur, OR Grade, Sklar®
Bone Cutters and Rongeurs
Love-Gruenwald intervertebral disc rongeurs are made of stainless steel.
  • With industrial standard
  • OR-grade
They are used to cut and remove bone fragments in the spinal column, and are available in a variety of lengths and cut patterns.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty