Would you like to log in again?
DeBakey Atraumatic Vena Cava Clamp, OR Grade, Sklar
Supplier: Sklar
52-6614 52-6612
10197-066EA 831.9 USD InStock
10197-066 10196-486
DeBakey Atraumatic Vena Cava Clamp, OR Grade, Sklar
Forceps
The DeBakey Atraumatic Vena Cava Clamp is a ratcheted, finger ring instrument, specially designed for total or partial occlusion of either the superior vena cava or the inferior vena cava. The inferior vena cava is the largest vein in the human body. Although it is a large vessel, it's walls are very thin and thus the benefit of using atraumatic clamps for controlling blood flow.
Sklar® operating room grade instruments are made from the highest grade surgical steel and incorporate the finest level of craftsmanship
Censitrac Ready products are industry standard and constructed of stainless steel.
Superior manufacturing and quality inspection guarantee that Sklar® instruments meet the demands of our customers. These instruments have been designed to provide exceptional comfort and precision all while ensuring the durability and strength that has been provided by Sklar for over 100 years. This product is considered a small size clamp.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty