Would you like to log in again?
DeBakey Atraumatic Tangential Occlusion Clamp, OR Grade, Sklar
Supplier: Sklar
52-6626 52-6622 52-6624 52-6628 52-6620
10197-076EA 876.31 USD InStock
10197-076 10197-074 10197-068 10197-072 10197-070
DeBakey Atraumatic Tangential Occlusion Clamp, OR Grade, Sklar
Forceps
DeBakey Atraumatic Tangential Occlusion Clamp is a ratcheted, finger ring clamp used in cardiovascular procedures to provide partial clamping of vessels.
The clamp holds a section of the vessel wall while blood continues to flow through the open section. The ratcheted handle allows the surgeon to adjust and maintain blood flow as necessary for the presenting conditions. The fine, nontraumatic teeth on this clamp causes little to no damage to the vessel. This clamp is also useful in vascular grafting procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty