Would you like to log in again?
DeBakey Atraumatic Multi-Purpose Clamps, OR Grade, Sklar
Supplier: Sklar
52-6645 52-6630 52-6631 52-6644 52-6640 52-6635 52-6634
10197-088EA 690.15 USD InStock
10197-088 10197-086 10196-240 10197-078 10197-080 10197-084 10197-082
DeBakey Atraumatic Multi-Purpose Clamps, OR Grade, Sklar
Forceps
DeBakey Atraumatic Multipurpose Clamps may be used as a hemostat to prevent blood flow, or as grasping forceps, since the atraumatic teeth provide a secure grip, but do not harm the tissues.
All forceps feature blunt tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty