Would you like to log in again?
Cooley Derra Anastomosis Clamp, OR Grade, Sklar
Supplier: Sklar
52-6956
10197-092EA 702.79 USD InStock
10197-092
Cooley Derra Anastomosis Clamp, OR Grade, Sklar
Forceps
Cooley-Derra Atraumatic Forceps are a ratcheted, finger ring instrument for anastomosis in cardiovascular procedures.
It is a partially occluding clamp used to control blood flow from the carotid artery, aorta, and inferior vena cava. The atraumatic teeth provide a secure but delicate grip on vessels, causing little to no damage to the vessel walls.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty