Would you like to log in again?
Sklar Black™ Gerald Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
90-1026 90-1025
10197-124EA 265.64 USD InStock
10197-124 10197-126
Sklar Black™ Gerald Dressing Forceps, OR Grade, Sklar
Forceps
Sklar Black™ Gerald Dressing Forceps are thumb style forceps commonly used for placing or removing dressings in many types of procedures
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed for laser surgeries.
This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
Although Dressing Forceps were designed to hold materials such as gauze. They may be used for removing foreign objects, or dead tissue from wounds, as well as other small surgical materials such as tubing. Serrated jaws assist in a having a secure hold on items.
Certifications: All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty