Would you like to log in again?
Parker Retractor, OR Grade, Sklar
Supplier: Sklar
60-1370 60-1350
10197-264PK 179.71 USD InStock
10197-264 10197-266
Parker Retractor, OR Grade, Sklar
Retractors
Sklar's Parker Double-Ended Retractor is a handheld retractor with two curved ends, sometimes refered to as "C" curves, with rounded tips
The Parker retractor is commonly used for holding back layers of surface or shallow tissue. They are typically used in pairs and as such, are sold in sets of two.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty