Would you like to log in again?
Farabeuf Retractor, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
60-1460 60-1461
10197-268PK 147.68 USD InStock
10197-268 10197-270
Farabeuf Retractor, OR Grade, Sklar
Retractors
Sklar Farabeuf Double-Ended Retractor is a versatile handheld retractor that is used in multiple procedures
It may be used in dentistry, wrist and hand procedures, or hernia repair, among other applications. The instrument is a solid piece of stainless steel with one angled blade at each end. Each blade is smooth, blunt, and also has a slight upward lip at the end.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty