Would you like to log in again?
Stevens Tenotomy Hook, OR Grade, Sklar®
Supplier: Sklar
65-3191
10197-346EA 126.79 USD InStock
10197-346
Stevens Tenotomy Hook, OR Grade, Sklar®
Retractors
This is often used for retracting and holding ocular muscles away from the surgical site during strabismus procedures.
  • Made of stainless steel
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty