Would you like to log in again?
Tyrell Iris Hook, OR Grade, Sklar
Supplier: Sklar
65-3206 65-3205
10197-352EA 51 USD InStock
10197-352 10197-354
Tyrell Iris Hook, OR Grade, Sklar
Retractors
The Sklar Tyrell Iris Hook is designed for retraction, dissection, handling, and the removal of delicate tissues.
It is most often used in ophthalmologic procedures but may also be used in dermatology procedures. One of the most popular skin hooks, the Tyrell Iris Hook is available with a blunt or sharp tip and has a smooth, flat handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty