Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Castroviejo Cyclodialysis Spatula, OR Grade, Sklar
Supplier: Sklar
65-4062 65-4072 65-4070 65-4066 65-4064
10197-384EA 266.13 USD InStock
10197-384 10197-382 75818-056 10197-378 10197-376
Castroviejo Cyclodialysis Spatula, OR Grade, Sklar
Spatulas
The Castroviejo cyclodialysis spatula is a double-ended ophthalmologic instrument commonly used in procedures for the treatment of glaucoma.
This double-ended product has a flat, straight, smooth handle that tapers gradually and extends out before the angled, spatula tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty