Would you like to log in again?
Williams Lacrimal Probe, OR Grade, Sklar
Supplier: Sklar
65-4125 65-4121 65-4120 65-4127 65-4123
10197-400EA 428.92 USD InStock
10197-400 10197-404 10197-402 10197-408 10197-406
Williams Lacrimal Probe, OR Grade, Sklar
Dissection Needles
Sklar's Williams Lachrimal Probe is a very thin malleable rod with a rounded end used in procedures within the lacrimal system, often pertaining to re-establishing tear flow.
This double-ended instrument has different size tips on each end and the center features a tongue tie that is used as a holding point. This device is available in a set of 4 which contains all sizes (1/2, 3/4, 5/6, and 7/8 probes), or in groups of two.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty