Would you like to log in again?
Castroviejo-Kalt Needle Holder, OR Grade, Sklar®
Supplier: Sklar
65-6055
10197-410EA 763.66 USD InStock
10197-410
Castroviejo-Kalt Needle Holder, OR Grade, Sklar®
Forceps
Sklar's® Castroviejo-Kalt Needle Holder is a multipurpose microsurgical instrument typically used during approximation and closure procedures
The handle of this needle holder varies from the double spring bow of the standard Castroviejo handle. The Castroviejo-Kalt handle is flat, but forms a triangular design with a thumb release. The jaws are still delicate and cross serrated. This product is straight with serrated tips and a length of 5-1/2".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty