Would you like to log in again?
Castroviejo Colibri Forceps, OR Grade, Sklar
Supplier: Sklar
66-3450
10197-496EA 855.32 USD InStock
10197-496
Castroviejo Colibri Forceps, OR Grade, Sklar
Forceps
Castroviejo Colibri Forceps are an ophthalmologic instrument used in cataract procedures.
They feature an extra delicate pattern for work with eye tissue. The toothed tip allows a more precise grip on the tissues being manipulated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty