Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
LiteGrip™ Iris Forceps, OR Grade, Sklar
Supplier: Sklar
66-3342 66-3542 66-3742
10197-494EA 155.24 USD InStock
10197-494 10197-498 75817-584
LiteGrip™ Iris Forceps, OR Grade, Sklar
Forceps
LiteGrip™ Iris Forceps are thumb forceps used for handling fine and delicate tissue and dressings during ophthalmic procedures.
LiteGrip™ instruments feature a lightweight, fenestrated handle. They are useful during lengthy procedures when fatigue may arise. The finger holes on the handle also allow grip adjustments as needed by the surgeon.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty