Would you like to log in again?
DeBakey Pean Forceps, OR Grade, Sklar
Supplier: Sklar
52-6840
10197-510EA 469.35 USD InStock
10197-510
DeBakey Pean Forceps, OR Grade, Sklar
Forceps
Sklar Debakey Pean Forceps are ratcheted, finger ring forceps used for grasping and clamping large vessels
They have 1x2 rows of fine, atraumatic teeth and cause little to no damage to the vessel walls. They may also be used for grasping heavy tissue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty