Would you like to log in again?
Gemini Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
55-2898 55-2890 55-2855
10197-582EA 288.51 USD InStock
10197-582 10197-580 10197-588
Gemini Forceps, OR Grade, Sklar
Forceps
Gemini Forceps, which may be referred to as Mixter forceps, are ratcheted, finger ring forceps used for clamping vessels and occluding tissue
Both Gemini and Mixter forceps have serrations the entire length of their jaws. Gemini forceps offer delicate serrations, so they may be used on delicate tissue. While Mixter forceps feature right angle jaws; Gemini tend to have more of a curve, both slight and full, to the jaws.
They are designed for vascular and cardiothoracic procedures.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty