Would you like to log in again?
Harrington Splanchnic Retractor, OR Grade, Sklar®
Supplier: Sklar
55-7125 55-7115
10197-612EA 691.8 USD InStock
10197-612 10197-610
Harrington Splanchnic Retractor, OR Grade, Sklar®
Retractors
Harrington Splanchnic Retractor is a handheld, manual retractor used to hold back tissue and organs deep in the abdominal region
  • The heart shaped tip of the blade is designed to reduce trauma to organs such as the liver and intestines. The handle has finger grips for a more secure hold and a hook at the bottom that can be used to rest the bottom finger and assist in the retraction process.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty