Would you like to log in again?
Burford-Finochietto Rib Spreader, OR Grade, Sklar
Supplier: Sklar
55-7980 55-7880
10197-650EA 3294.02 USD InStock
10197-650 10197-648
Burford-Finochietto Rib Spreader, OR Grade, Sklar
Retractors
The Burford-Finochietto Rib Spreader features two sets of blades (Large: 2.5×2.5" and Small: 1.5×2.5"), with an 8" spread.
  • Industry standard
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty