Would you like to log in again?
Metzenbaum-Nelson Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
55-9184 55-9180 55-9084 55-9080
10197-670EA 245.02 USD InStock
10197-670 10197-674 10193-906 10197-676
Metzenbaum-Nelson Dissecting Scissors, OR Grade, Sklar®
Scissors
Metzenbaum-Nelson Dissecting Scissors are used for dissecting and cutting tissue.
These scissors are not recommended for cutting sutures, drains, or heavy tissue
Censitrac Ready products are industry standard and constructed of stainless steel.
Metzenbaum-Nelson scissors tend to be longer than standard Metzenbaum scissors and are ideal for use in thoracic procedures.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty