Would you like to log in again?
Metzenbaum-Nelson Bariatric Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
55-9614 55-9612
10197-686EA 442.24 USD InStock
10197-686 10197-688
Metzenbaum-Nelson Bariatric Dissecting Scissors, OR Grade, Sklar
Scissors
Metzenbaum-Nelson Dissecting Scissors are designed for dissecting and cutting tissue.
They tend to be longer than standard Metzenbaum scissors, especially for use in thoracic procedures.
Not recommended for cutting sutures, drains, or heavy tissue.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty