Would you like to log in again?
Green Retractor, OR Grade, Sklar
Supplier: Sklar
57-1485
10197-698EA 181.25 USD InStock
10197-698
Green Retractor, OR Grade, Sklar
Retractors
Sklar's Green Retractor is a handheld, single-end retractor most commonly used in thyroidectomy and other procedures in the neck
It has a strongly curved, smooth, fenestrated blade. It also has a teardrop fenestrated handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty