Would you like to log in again?
Goelet Retractor, OR Grade, Sklar
Supplier: Sklar
60-1075
10197-700EA 180.9 USD InStock
10197-700
Goelet Retractor, OR Grade, Sklar
Retractors
Sklar's Goelet Double-Ended Retractor is a handheld retractor used for holding back tissue from small, superficial incisions
This retractor has strongly curved, saddle-style shaped blades.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty