Would you like to log in again?
Little Retractor with Extra Thin Blade, OR Grade, Sklar®
Supplier: Sklar
60-5472
10197-776EA 260.65 USD InStock
10197-776
Little Retractor with Extra Thin Blade, OR Grade, Sklar®
Retractors
Sklar's® Little Retractor is a single-end, handheld instrument used to hold back intermediate depth tissue
The ½" wide blade is smooth with a rounded tip and strong curve. The Little Retractor features a flat, grooved handle.
Ordering information: Each sold separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty