Would you like to log in again?
Mathieu Retractor, OR Grade, Sklar
Supplier: Sklar
60-6062 60-6064
10197-808EA 242.89 USD InStock
10197-808 10194-440
Mathieu Retractor, OR Grade, Sklar
Retractors
Sklar's Mathieu Double-Ended Retractor is a handheld instrument used to hold back muscle and tissue close to the surface
The ends of the Mathieu retractor angle out in the same direction. One end is forked with three curved prongs and the other is a solid, flat blade angled at approximately 90°. The handle of the retractor is flat and smooth. This product features blunt tips on the forked end of the retractor. The Mathieu retractor is almost identical to the Senn retractor, however the ends of the Senn retractor angle out in opposite directions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty