Would you like to log in again?
Enucleation Scissors, OR Grade, Sklar
Supplier: Sklar
64-1150
10197-846EA 235.87 USD InStock
10197-846
Enucleation Scissors, OR Grade, Sklar
Scissors
Enucleation Scissors are an ophthalmic instrument used primarily in procedures dealing with removal of the globe due to tumor or trauma
These finger ring scissors have thick blades that allow the surgeon to manuever around the globe and reach the optic nerve in order to sever it. The edges are smooth and the tips are blunt.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty