Would you like to log in again?
Castroviejo Speculum, OR Grade, Sklar®
Supplier: Sklar
65-1535
10197-912EA 777.51 USD InStock
10197-912
Castroviejo Speculum, OR Grade, Sklar®
Specula
Sklar's® Castroviejo Eye Speculum is a self-retaining retractor used in ophthalmic procedures.
  • Designed with 15×5 mm fenestrated blades
There is a locking mechanism located at the base of the instrument, and it is available in three sizes. The instrument is slightly curved to adapt to the curvature of the patient's face.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty