Would you like to log in again?
Kristeller Vaginal Speculum and Retractor Set, OR Grade, Sklar
Supplier: Sklar
90-3950
10197-950EA 297.55 USD InStock
10197-950
Kristeller Vaginal Speculum and Retractor Set, OR Grade, Sklar
Specula
Kristeller Vaginal Speculum and Retractor Set are used in OB/GYN procedures.
This set includes a 90×36 mm speculum and a 100×32 mm retractor. The sides of the speculum blade are curved upward slightly. The retractor has a smooth, flat blade. Both feature a slight hook at the bottom of the handle for pulling.
They are used for visual examinations because both are handheld, manual, non-locking instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty