Would you like to log in again?
Kogan Endocervical Speculum, OR Grade, Sklar
Supplier: Sklar
90-3995 90-3996 90-3997 90-3998 90-3990 90-3994
10197-960EA 837.7 USD InStock
10197-960 10193-356 10197-954 10197-958 10193-338 10197-956
Kogan Endocervical Speculum, OR Grade, Sklar
Specula
Sklar's kogan endocervical speculum is a gynecological instrument used for viewing the distal endocervical canal.
  • Ratchet locking mechanism
  • With narrow blades of 2 to 4 mm wide
  • Serrations outside of the blade
Unlike a traditional duckbill speculum, this single-spring, locking, finger-ring intrument features opposing, fenestrated blades at the end of long jaws. The jaws and blades extend deeper into the cervix and can therefore retract deeper tissue.
An endocervical speculum is frequently used in colposcopy procedures, or where polyps cannot be easily visualized.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty