Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Phos-tag™ Acrylamide, Wako
Supplier: Wako
304-93526 304-93521 300-93523
101974-090EA 313.85 USD InStock
101974-090 101974-086 101974-088
Phos-tag™ Acrylamide, Wako
Electrophoresis Reagents Western Blotting Reagents
Phos-tag™ is a novel phosphate-binding tag at neutral pH.
  • Ideal for separation
  • Purifies and enriches phosphorylated substances
It is ideal for separation, purification and enrichment of phosphorylated substances. Phosphorylated proteins and non-phosphorylated proteins were inseparable with the conventional SDS-PAGE, in the absence of Phos-tag™.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty