Would you like to log in again?
Bishop-Harmon Micro Iris Forceps, OR Grade, Sklar
Supplier: Sklar
66-4234 66-4232
10198-010EA 296.96 USD InStock
10198-010 10194-584
Bishop-Harmon Micro Iris Forceps, OR Grade, Sklar
Forceps
Sklar Bishop-Harmon Ophthalmic Tissue Forceps are primarily used in ophthalmic procedures or microsurgery
They are delicate forceps with 1x2 fine teeth. Standard Bishop-Harmon forceps have fenestrated handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty