Would you like to log in again?
Foerster Iris Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
66-4530
10198-012EA 188.58 USD InStock
10198-012
Foerster Iris Tissue Forceps, OR Grade, Sklar
Forceps
Foerster Iris Tissue Forceps are thumb forceps used for handing fine, delicate tissue, often during ophthalmic procedures
Tissue Forceps, unlike Dressing Forceps, are toothed so they can grip tissue, which may be slippery. The handle features a fenestrated octagonal thumb/finger grip for increased precision, control, and comfort.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty