Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Arruga Capsule Forceps, OR Grade, Sklar
Supplier: Sklar
66-5340
10198-022EA 466.07 USD InStock
10198-022
Arruga Capsule Forceps, OR Grade, Sklar
Forceps
Arruga Capsule Forceps are an ophthalmic instrument used for holding the lens capsule during cataract procedures.
The tips are designed not to damage the iris. They are oval shaped with a slight cup and a standard curve.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty