Would you like to log in again?
Castroviejo Suture Forceps, OR Grade, Sklar
Supplier: Sklar
66-6116 66-6105 66-6115 66-6118 66-6109
10198-040EA 765.39 USD InStock
10198-040 94002-756 10198-032 10198-042 10198-038
Castroviejo Suture Forceps, OR Grade, Sklar
Forceps
Castroviejo Suture Forceps are a versatile microsurgical instrument used in ophthalmologic and plastic procedures near the eyes.
The forceps are used to hold both tissue and suture materials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty