Would you like to log in again?
McPherson Tying Forceps, OR Grade, Sklar
Supplier: Sklar
66-6150 66-6152
10198-046EA 425.06 USD InStock
10198-046 10198-048
McPherson Tying Forceps, OR Grade, Sklar
Forceps
McPherson Tying Forceps are an ophthalmologic instrument used for suturing
The fine tips feature a smooth tying platform for use in cataract procedures.
They should not be used for grasping tissue due to the potential damage that may result.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty