Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Gill (Chandler) Iris Forceps, OR Grade, Sklar
Supplier: Sklar
66-7244
10198-090EA 441.25 USD InStock
10198-090
Gill (Chandler) Iris Forceps, OR Grade, Sklar
Forceps
Gill (Chandler) Iris Dressing Forceps are thumb style forceps, used for placing or removing dressings in many procedures.
Although Dressing Forceps were designed to hold materials such as gauze, they have a variety of additional functions. They may be used for removing foreign objects or dead tissue from wounds, as well as other small surgical materials such as tubing. Serrated jaws assist in a having a secure hold on items.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty