Would you like to log in again?
Hartmann Alligator Forceps, OR Grade, Sklar
Supplier: Sklar
67-1030
10198-112EA 1281.77 USD InStock
10198-112
Hartmann Alligator Forceps, OR Grade, Sklar
Forceps
Sklar Hartmann Alligator Forceps are an ENT instrument commonly used in myringotomy procedures to insert or remove tubes or dressings and packing
The serrated tips securely hold and remove foreign objects, granulation tissue, and cerumen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty