Would you like to log in again?
Jansen Bayonet Nasal Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
67-1465 70-4375
10198-284EA 101 USD InStock
10198-284 10194-430
Jansen Bayonet Nasal Forceps, OR Grade, Sklar
Forceps
Sklar® Jansen Bayonet Dressing Forceps are thumb forceps commonly used in nasal procedures for placing packing.
The bayonet style handle and long jaws allow an unobstructed view when working in the nasal cavity. Serrated tips securely hold surgical materials. These forceps may also be used in intracranial procedures. This product has straight, serrated tips.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty