Would you like to log in again?
Alexander Mastoid Chisel, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
67-4608 67-4612 67-4614 67-4606 67-4610 67-4604
10198-382EA 203.18 USD InStock
10198-382 10198-384 75817-956 75817-958 10198-378 10198-380
Alexander Mastoid Chisel, OR Grade, Sklar
Chisels
The Sklar Alexander mastoid chisel is similar to the Alexander mastoid gouge but does not have the scoop-style end.
The chisel is a solid piece of stainless steel with one blunt end for hammering and a sharp, chiseled tip for removing bone at the opposite end.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty