Would you like to log in again?
Blumenthal Rongeur, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
67-5994 67-5990
10198-404EA 528.04 USD InStock
10198-404 10198-406
Blumenthal Rongeur, OR Grade, Sklar®
Bone Cutters and Rongeurs
Sklar's® Blumenthal Rongeur is a double spring, plier instrument used in a variety of procedures to trim and remove bone.
  • Made of stainless steel
While large amounts of bone can be removed with rongeurs, pieces should be removed in small bites. Blumenthal rongeurs are available with three different angle tips, which allows them to be both side and end cutting and therefore useful in difficult to reach areas. They are often used in dental procedures to remove inter-radicular bone.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty