Would you like to log in again?
Otoscope, OR Grade, Sklar
Supplier: Sklar
67-6250
10198-452EA 283.69 USD InStock
10198-452
Otoscope, OR Grade, Sklar
Opthalmoscopes
The Sklar Otoscope employs a Rheostat for varied light intensity.
  • Rheostat provided for varied light intensity
  • Three specula
  • Chrome plated
  • Reusable
  • Latex-free
This Otoscope has three specula (sizes 2.5 mm, 3.5 mm, 4.5 mm).
Ordering information: Batteries required, need to be ordered separatly.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty