Would you like to log in again?
Micro Alligator Ear Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
67-8178 67-8180 67-8010 67-8008 67-8006 67-8170 67-8176 67-8177 67-8172 67-8024 67-8174 67-8000
10198-472EA 717.06 USD InStock
10198-472 10193-944 10198-482 10192-462 10198-462 10198-484 10194-260 10198-464 10198-486 10198-488 10193-772 10198-490
Micro Alligator Ear Forceps, OR Grade, Sklar
Forceps
Alligator Ear Forceps are finger ring ENT forceps used for manipulating and removing tissue from the ear canal or middle ear
They may also be used for packing during grafting procedures or as nasal forceps.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty