Would you like to log in again?
Littauer Nasal Forceps, OR Grade, Sklar
Supplier: Sklar
70-4170
10198-530EA 559.06 USD InStock
10198-530
Littauer Nasal Forceps, OR Grade, Sklar
Forceps
Littauer Nasal Forceps are a finger ring instrument used in ear and nasal procedures.
This product has short shanks that angle at the screw lock to a 4 1/2" long shaft. The jaws are useful for grasping at tissue or foreign objects, that may not be easily seen or reached in the ear or nasal canal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty