Would you like to log in again?
Noyes Alligator Forceps, OR Grade, Sklar
Supplier: Sklar
70-5235 70-5332 70-5232 70-5336 67-1734 70-5335
10198-560EA 757.3 USD InStock
10198-560 10198-286 10198-568 10193-310 10198-138 10198-558
Noyes Alligator Forceps, OR Grade, Sklar
Forceps
Noyes Alligator Forceps are a finger ring instrument used for ENT procedures.
The long shaft allows access to deep, narrow spaces such as the ear and nasal canals. Serrated jaws provide additional grip for removing foreign objects. This product has straight 3" shanks that angle into a 5 1/2" shaft at the box lock. The tips are serrated with 1 x 2 teeth and a central groove as well.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty