Would you like to log in again?
Barnhill Adenoid Curette, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
70-9200 70-9203 70-9201 70-9202 70-9204
10198-570EA 431.53 USD InStock
10198-570 10198-572 10198-574 10198-576 10198-578
Barnhill Adenoid Curette, OR Grade, Sklar
Curettes
Barnhill adenoid curette is a debulking instrument used primarily in adenoidectomy procedures.
  • Curved and rectangular
  • Fenestrated tip
The Barnhill Adenoid Curette is similar to the Beckman adenoid curette; however, the working end of the Beckman curette is slightly wider and shorter, resembling more of a square shape. It is also slightly longer in overall length.
It has a sharp inner blade that is used to scrape away adenoids or tonsils. It is available with a variety of blade widths.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty