Would you like to log in again?
Beckman Adenoid Curette, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
70-9222 70-9220 70-9221 70-9223
10198-582EA 374.72 USD InStock
10198-582 10198-584 10198-586 10198-588
Beckman Adenoid Curette, OR Grade, Sklar®
Curettes
Sklar's® Beckman Adenoid Curette is a debulking instrument used primarily in adenoidectomy procedures.
  • Curved, square, fenestrated tip with a sharp inner blade that is used to scrape away tissue following removal of the adenoids or tonsils
  • Available with a variety of blade widths
This instrument is similar to the Barnhill adenoid curette; however, the working end of the Barnhill curette is slightly longer and more narrow, resembling more of a rectangular shape. The Barnhill curette is also slightly shorter in overall length.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty