Would you like to log in again?
Kelly Hemostatic Forceps, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
74-3270 17-2055 74-3260 17-2155 18-1552 18-1652
82029-976EA 67.77 USD InStock
82029-976 10192-370 10185-272 10198-616 82029-974 10185-270
Kelly Hemostatic Forceps, OR Grade, Sklar
Forceps
Kelly Hemostatic Forceps are primarily used for clamping large blood vessels or manipulating heavy tissue
  • Available curved or straight
  • With a ratcheted finger ring handle
  • Latex-free
They may also be used for soft tissue dissection. The jaws of Kelly Forceps are 1/3 the length of the shanks, and serrations are 1/2 the length of the jaws.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty