Would you like to log in again?
Hurd Tonsil Dissector and Pillar Retractor, OR Grade, Sklar
Supplier: Sklar
75-6387
10198-660EA 212.33 USD InStock
10198-660
Hurd Tonsil Dissector and Pillar Retractor, OR Grade, Sklar
Retractors
Sklar's Hurd Tonsil Dissector and Pillar Retractor is an ENT instrument used in oral procedures and tonsillectomies
It is a double-ended instrument with a smooth retractor lip end as well as a slightly sharp, rounded dissecting end.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty