Would you like to log in again?
Snare Wires, OR Grade, Sklar
Supplier: Sklar
75-9508 75-9408 67-1753 75-9507 75-9407 67-1752 70-6168 75-9406 75-9405 75-9509 75-9409 75-9505 75-9506
10198-690EA 23.38 USD InStock
10198-690 10198-692 10198-680 10198-682 10198-154 10198-684 10198-686 10198-678 10161-924 10198-688 10163-402 10198-290 10198-292
Snare Wires, OR Grade, Sklar
Wire
Snare Wire may be used in tonsil, nasal, or ear snare instruments commonly used for removing polyps and other growths in difficult to reach cavity areas.
The instrument itself controls the wire, which loops around the base of the mass to be removed. The instrument tightens the wire which removes the growth. Snare Wires are constructed of stainless steel. Industry standard.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty